• USA Home
  • 02463 - 17α-Ethynylestradiol

EMAIL THIS PAGE TO A FRIEND
02463 Sigma-Aldrich

17α-Ethynylestradiol

≥98.0% (HPLC)

Synonym: 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol

  • CAS Number 57-63-6

  • Empirical Formula (Hill Notation) C20H24O2

  • Molecular Weight 296.40

  •  Beilstein/REAXYS Number 2419975

  •  EC Number 200-342-2

  •  MDL number MFCD00003690

  •  PubChem Substance ID 329747438

Purchase

Properties

Related Categories Biochemicals and Reagents, Estrogens (C18) and Derivatives, Lipids, Sterol Lipids
InChI Key   BFPYWIDHMRZLRN-SLHNCBLASA-N
assay   ≥98.0% (HPLC)
mp   182-183 °C (lit.)
solubility   ethanol: 50 mg/mL, clear, slightly yellow

Description

Other Notes

Affects lipoprotein lipase and hepatic lipase activities, consequently used in studies of VLDL metabolism

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
RC8925000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?