EMAIL THIS PAGE TO A FRIEND
02940 Sigma-Aldrich

Ethylamine

purum, anhydrous, ≥99.0%

Synonym: Aminoethane, Monoethylamine

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
vapor density   1.56 (15 °C, vs air)
vapor pressure   874 mmHg ( 20 °C)
grade   purum
assay   ≥99.0%
autoignition temp.   721 °F
quality   anhydrous
expl. lim.   14 %
  3.5-14 %
impurities   ≤0.2% NH3
  ≤0.5% (CH3)2NH
bp   16.6 °C (lit.)
mp   -81 °C (lit.)
density   0.680 g/mL at 20 °C (lit.)
  0.689 g/mL at 25 °C (lit.)
SMILES string   CCN
InChI   1S/C2H7N/c1-2-3/h2-3H2,1H3
InChI key   QUSNBJAOOMFDIB-UHFFFAOYSA-N

Description

Packaging

pressure tin with 100 mL (net ~68 g)

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1036 - class 2 - Ethylamine
WGK Germany 
1
RTECS 
KH2100000
Flash Point(F) 
-34.6 °F
Flash Point(C) 
-37 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?