• USA Home
  • 03734 - Europium Standard for AAS
EMAIL THIS PAGE TO A FRIEND
03734 Sigma-Aldrich
Europium Standard for AAS
TraceCERT®, 1000 mg/L Eu in nitric acid
Purchase
Description
Analysis Note
1000 mg/L Eu in 2% nitric acid, prepared with high purity Eu2O3, HNO3 and water
General description
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Other Notes
For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website
Legal Information
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
1
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?