EMAIL THIS PAGE TO A FRIEND
05022 Supelco

N,N-Dimethylglycine

for HPLC derivatization, LiChropur, ≥99% (NT)

Synonym: (Dimethylamino)acetic acid

  • CAS Number 1118-68-9

  • Linear Formula (CH3)2NCH2COOH

  • Molecular Weight 103.12

  •  Beilstein/REAXYS Number 1700261

  •  EC Number 214-267-8

  •  MDL number MFCD00004283

  •  PubChem Substance ID 329748006

  •  NACRES NA.22

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents HPLC, Mass Spectrometry More...
Quality Level   100
grade   for HPLC derivatization
assay   ≥99% (NT)
quality   LiChropur
application(s)   HPLC: suitable
mp   178-182 °C (lit.)
suitability   suitable for LC-MS
SMILES string   CN(C)CC(O)=O
InChI   1S/C4H9NO2/c1-5(2)3-4(6)7/h3H2,1-2H3,(H,6,7)
InChI key   FFDGPVCHZBVARC-UHFFFAOYSA-N

Description

General description

N,N-Dimethylglycine is a type of quaternary ammonium compound which exhibits a variety of biological effects.

Application

N,N-Dimethylglycine has been used as a reference standard for the quantification of N,N-dimethylglycine in human plasma and serum by normal phase high performance liquid chromatography combined with triple-quadrupole tandem mass spectrometer (HPLC-MS/MS) operating on positive-ion mode, in human urine by stable-isotope dilution ultra-performance liquid chromatography−tandem mass spectrometry (UPLC−MS/MS) as well as UPLC coupled with quadrupole linear ion trap MS and in various soy sauces as hydrophilic metabolites by HPLC-MS.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
MB9865000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?