• USA Home
  • 05392 - 2-Methyl-3-buten-2-ol

EMAIL THIS PAGE TO A FRIEND
05392 Supelco

2-Methyl-3-buten-2-ol

analytical standard

Synonym: 1,1-Dimethylallyl alcohol, 3-Hydroxy-3-methyl-1-butene

  • CAS Number 115-18-4

  • Linear Formula CH2=CHC(CH3)2OH

  • Molecular Weight 86.13

  •  Beilstein/REAXYS Number 1698263

  •  EC Number 204-068-4

  •  MDL number MFCD00004470

  •  PubChem Substance ID 329748056

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alphabetical Index of Phytochemical Standards, Analytical Standards,
Quality Level   100
grade   analytical standard
vapor pressure   51 mmHg ( 25 °C)
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.416 (lit.)
bp   98-99 °C (lit.)
density   0.824 g/mL at 25 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
storage temp.   2-8°C
SMILES string   CC(C)(O)C=C
InChI   1S/C5H10O/c1-4-5(2,3)6/h4,6H,1H2,2-3H3
InChI key   HNVRRHSXBLFLIG-UHFFFAOYSA-N

Description

General description

2-Methyl-3-buten-2-ol (MBO) is a biogenic, 5-carbon volatile alcohol released into the atmosphere by pine trees. It is also an important component of the bark beetle (Ips typographus) pheromone. MBO is considered to be a potential biofuel due to its attractive properties, including high energy content and low solubility in water.

Application

2-Methyl-3-buten-2-ol may be used as an analytical reference standard for the determination of the analyte in Humulus Lupulus L. (Hop) and pine-mushroom extracts by chromatography techniques.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1987BE 3 / PGII
WGK Germany 
1
RTECS 
EM9472000
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?