EMAIL THIS PAGE TO A FRIEND
05675 Sigma-Aldrich

Allene

purum, ≥96.0%

Synonym: Propadiene

Purchase

Properties

vapor density   1.42 (20 °C, vs air)
vapor pressure   6795 mmHg ( 21 °C)
grade   purum
assay   ≥96.0%
expl. lim.   13 %
bp   −34 °C (lit.)
mp   −136 °C (lit.)
storage temp.   2-8°C
SMILES string   C=C=C
InChI   1S/C3H4/c1-3-2/h1-2H2
InChI key   IYABWNGZIDDRAK-UHFFFAOYSA-N

Description

Packaging

lecture bottle

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2200 2.1
WGK Germany 
3
Protocols & Articles

Articles

GC Analysis of C1-C5 Hydrocarbons on Alumina Sulfate PLOT

From our library of Articles, Sigma-Aldrich presents GC Analysis of C1-C5 Hydrocarbons on Alumina Sulfate PLOT
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Sample preparations

GC Analysis of Impurities in Propylene on Alumina Chloride PLOT

From our library of Articles, Sigma-Aldrich presents GC Analysis of Impurities in Propylene on Alumina Chloride PLOT
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?