• USA Home
  • 05700 - 2,6-Dimethyl-2,4,6-octatriene

EMAIL THIS PAGE TO A FRIEND
05700 Sigma-Aldrich

2,6-Dimethyl-2,4,6-octatriene

technical, mixture of isomers, ~80% (GC)

Synonym: Alloocimene

  • CAS Number 673-84-7

  • Linear Formula CH3CH=C(CH3)CH=CHCH=C(CH3)2

  • Molecular Weight 136.23

  •  Beilstein Registry Number 1719993

  •  EC Number 211-614-5

  •  MDL number MFCD00009278

  •  PubChem Substance ID 24845866

Purchase

Properties

InChI Key   GQVMHMFBVWSSPF-SOYUKNQTSA-N
grade   technical
assay   ~80% (GC)
refractive index   n20/D 1.542(lit.)
  n20/D 1.543
bp   73-75 °C/14 mmHg(lit.)
density   0.811 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
3
Flash Point(F) 
156.2 °F
Flash Point(C) 
69 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

289698 2,6-Dimethyl-2,4,6-octatriene, technical grade, 80%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?