• USA Home
  • 05787 - Phosphorus(V) oxybromide

EMAIL THIS PAGE TO A FRIEND
05787 Sigma-Aldrich

Phosphorus(V) oxybromide

puriss., ≥98.5% (AT)

Synonym: Phosphorus oxide bromide, Phosphoryl bromide

Purchase

Properties

Related Categories Bromination, C-X Bond Formation (Halogen), Chemical Synthesis, Synthetic Reagents
InChI Key   UXCDUFKZSUBXGM-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (AT)
bp   192 °C(lit.)
mp   56 °C (lit.)
density   2.82 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 1939 8 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

376949 Phosphorus(V) oxybromide, >95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?