EMAIL THIS PAGE TO A FRIEND
05808 Sigma-Aldrich

Allylamine

puriss., ≥99.5% (GC)

Synonym: 2-Propen-1-ylamine, 3-Amino-1-propene

  • CAS Number 107-11-9

  • Linear Formula CH2=CHCH2NH2

  • Molecular Weight 57.09

  •  Beilstein/REAXYS Number 635703

  •  EC Number 203-463-9

  •  MDL number MFCD00008199

  •  PubChem Substance ID 329748171

  •  NACRES NA.22

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   2 (vs air)
vapor pressure   4.09 psi ( 20 °C)
grade   puriss.
assay   ≥99.5% (GC)
expl. lim.   22 %
impurities   ≤0.2% water
refractive index   n20/D 1.420 (lit.)
  n20/D 1.420
bp   53 °C (lit.)
mp   −88 °C (lit.)
density   0.761 g/mL at 25 °C (lit.)
SMILES string   NCC=C
InChI   1S/C3H7N/c1-2-3-4/h2H,1,3-4H2
InChI key   VVJKKWFAADXIJK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2334 3(6.1) / PGI
WGK Germany 
2
RTECS 
BA5425000
Flash Point(F) 
-18.4 °F
Flash Point(C) 
-28 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

241075 Allylamine, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?