• USA Home
  • 06251 - Aluminum isopropoxide

EMAIL THIS PAGE TO A FRIEND
06251 Sigma-Aldrich

Aluminum isopropoxide

purum, dist., ≥97.0% (KT)

Synonym: AIP, Aluminum isopropylate, Aluminum triisopropoxide

  • CAS Number 555-31-7

  • Linear Formula Al[OCH(CH3)2]3

  • Molecular Weight 204.24

  •  Beilstein/REAXYS Number 3910275

  •  EC Number 209-090-8

  •  MDL number MFCD00008870

  •  PubChem Substance ID 329748226

Purchase

Properties

Related Categories Acids, Acids & Bases, Aluminum, Aluminum Catalysts, Catalysis and Inorganic Chemistry,
InChI Key   SMZOGRDCAXLAAR-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (KT)
form   solid
quality   dist.
mp   128-133 °C (lit.)
  134-138 °C
density   1.035 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3181 4.1 / PGII
WGK Germany 
1
RTECS 
BD0975000
Flash Point(F) 
60.8 °F
Flash Point(C) 
16 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?