EMAIL THIS PAGE TO A FRIEND
06568 Sigma-Aldrich

Amikacin dihydrate

≥98.0% (TLC)

Synonym: N1-[(S)-4-Amino-2-hydroxybutyryl]-kanamycin A

  • CAS Number 37517-28-5

  • Empirical Formula (Hill Notation) C22H43N5O13 · 2H2O

  • Molecular Weight 621.63

  •  Beilstein Registry Number 1445422

  •  EC Number 253-538-5

  •  MDL number MFCD00151224

Purchase

Properties

assay   ≥98.0% (TLC)
optical activity   [α]20/D +93±2°, c = 2% in H2O
solubility   H2O: 50 mg/mL, clear, colorless
storage temp.   2-8°C

Description

Other Notes

Aminoglycoside antibiotic

General description

Chemical structure: aminoglycoside

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
WK1955000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A3650 Amikacin hydrate, aminoglycoside antibiotic

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?