• USA Home
  • 07549 - (Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
07549 Supelco

(Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate

≥97.0% (TLC)

Synonym: PyBOP®

  • CAS Number 128625-52-5

  • Linear Formula C18H28N6OP · PF6

  • Molecular Weight 520.39

  •  Beilstein/REAXYS Number 3584612

  •  MDL number MFCD00077411

  •  PubChem Substance ID 329748476

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Coupling, Peptide Coupling, Peptide Synthesis and Peptide Chemistry,
assay   ≥97.0% (TLC)
mp   ~150 °C
  154-156 °C (dec.) (lit.)
SMILES string   F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](On2nnc3ccccc23)(N4CCCC4)N5CCCC5
InChI   1S/C18H28N6OP.F6P/c1-2-10-18-17(9-1)19-20-24(18)25-26(21-11-3-4-12-21,22-13-5-6-14-22)23-15-7-8-16-23;1-7(2,3,4,5)6/h1-2,9-10H,3-8,11-16H2;/q+1;-1
InChI key   VIAFLMPQBHAMLI-UHFFFAOYSA-N

Description

Application

Analogue of the BOP coupling reagent which does not form carcinogenic HMPA as by-product.

Legal Information

PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

COMU – Safer and More Efficient Peptide Coupling Reagent

Peptide synthesis relies heavily on efficient and reliable coupling reagents. A low tendency for racemization is a key requirement. This is especially true for solid phase peptide synthesis—quantitat...
Keywords: Building blocks, Calorimetry, Condensations, Coupling reactions, Epimerizations, PEGylations, Peptide synthesis, Racemizations, Solid phase peptide synthesis, Solvents

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

12805 (Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, purum, ≥97.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?