EMAIL THIS PAGE TO A FRIEND
07621 Sigma-Aldrich

Cycloleucine

purum, ≥97.0% (NT)

Synonym: 1-Aminocyclopentanecarboxylic acid

  • CAS Number 52-52-8

  • Linear Formula H2NC5H8CO2H

  • Molecular Weight 129.16

  •  Beilstein Registry Number 636626

  •  EC Number 200-144-6

  •  MDL number MFCD00001381

  •  PubChem Substance ID 24846087

Purchase

Properties

InChI Key   NILQLFBWTXNUOE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (NT)
mp   320 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
GY2625000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?