• USA Home
  • 07665 - N,N-Diethyl-p-phenylenediamine oxalate salt

EMAIL THIS PAGE TO A FRIEND
07665 Sigma-Aldrich

N,N-Diethyl-p-phenylenediamine oxalate salt

technical, ≥90% (T)

Synonym: 4-Amino-N,N-diethylaniline oxalate salt

  • CAS Number 142439-89-2

  • Empirical Formula (Hill Notation) C10H16N2 · C2H2O4

  • Molecular Weight 254.28

  •  MDL number MFCD00042025

  •  PubChem Substance ID 329748499

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
InChI Key   GXSUUFAGHVDMCO-UHFFFAOYSA-N
grade   technical
assay   ≥90% (T)
impurities   ≤5% water
  0.5-1 mol/mol oxalic acid
mp   145 °C (dec.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?