EMAIL THIS PAGE TO A FRIEND
08053 Sigma-Aldrich

6-Amino-1-hexanol

purum, ≥97.0% (NT)

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
grade   purum
assay   ≥97.0% (NT)
bp   135-140 °C/30 mmHg (lit.)
mp   54-58 °C (lit.)
  54-58 °C
SMILES string   NCCCCCCO
InChI   1S/C6H15NO/c7-5-3-1-2-4-6-8/h8H,1-7H2
InChI key   SUTWPJHCRAITLU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MO8840000

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?