• USA Home
  • 08060 - N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
08060 Sigma-Aldrich

N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride

≥99.0% (TLC)

Synonym: W-7

  • CAS Number 61714-27-0

  • Empirical Formula (Hill Notation) C16H21ClN2O2S · HCl

  • Molecular Weight 377.33

  •  Beilstein/REAXYS Number 6030174

  •  MDL number MFCD00012559

  •  PubChem Substance ID 329748545

Purchase

Properties

InChI Key   OMMOSRLIFSCDBL-UHFFFAOYSA-N
assay   ≥99.0% (TLC)
impurities   ≤1.0% diaminohexane (TLC)
mp   220-222 °C
solubility   methanol: 25 mg/mL, clear, colorless
Mode of action   enzyme | inhibits
storage temp.   −20°C

Description

Other Notes

Calmodulin antagonist, review

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?