• USA Home
  • 08396 - (S)-(−)-1-Amino-2-(methoxymethyl)pyrrolidine

EMAIL THIS PAGE TO A FRIEND
08396 Sigma-Aldrich

(S)-(−)-1-Amino-2-(methoxymethyl)pyrrolidine

purum, ≥95.0% (sum of enantiomers, GC)

Synonym: SAMP

  • CAS Number 59983-39-0

  • Empirical Formula (Hill Notation) C6H14N2O

  • Molecular Weight 130.19

  •  Beilstein Registry Number 1523794

  •  MDL number MFCD00064485

  •  PubChem Substance ID 24846168

Purchase

Properties

InChI Key   BWSIKGOGLDNQBZ-LURJTMIESA-N
grade   purum
assay   ≥95.0% (sum of enantiomers, GC)
optical activity   [α]20/D −82±2°, neat
refractive index   n20/D 1.465(lit.)
  n20/D 1.465
bp   42 °C/1.8 mmHg(lit.)
density   0.97 g/mL at 25 °C(lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?