• USA Home
  • 08400 - 4-(Aminomethyl)benzoic acid

EMAIL THIS PAGE TO A FRIEND
08400 Sigma-Aldrich

4-(Aminomethyl)benzoic acid

purum, ≥96.0% (calc. based on dry substance, NT)

  • CAS Number 56-91-7

  • Linear Formula H2NCH2C6H4CO2H

  • Molecular Weight 151.16

  •  Beilstein Registry Number 1100606

  •  EC Number 200-297-9

  •  MDL number MFCD00010203

  •  PubChem Substance ID 24846170

Purchase

Properties

InChI Key   QCTBMLYLENLHLA-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (calc. based on dry substance, NT)
impurities   ~4% water
mp   ≥300 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

283746 4-(Aminomethyl)benzoic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?