• USA Home
  • 08990 - 2-Methyl-4-nitroaniline

EMAIL THIS PAGE TO A FRIEND
08990 Sigma-Aldrich

2-Methyl-4-nitroaniline

purum, ≥98.0% (HPLC)

Synonym: 2-Amino-5-nitrotoluene, 4-Nitro-o-toluidine

  • CAS Number 99-52-5

  • Linear Formula CH3C6H3(NO2)NH2

  • Molecular Weight 152.15

  •  Beilstein Registry Number 775772

  •  EC Number 202-762-1

  •  MDL number MFCD00007734

  •  PubChem Substance ID 24846241

Purchase

Properties

InChI Key   XTTIQGSLJBWVIV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   130-132 °C (lit.)
  130-132 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2660 6.1 / PGIII
WGK Germany 
3
RTECS 
XU8210000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?