• USA Home
  • 08991 - 2-Methyl-3-nitroaniline

EMAIL THIS PAGE TO A FRIEND
08991 Sigma-Aldrich

2-Methyl-3-nitroaniline

purum, ≥98.0% (HPLC)

Synonym: 2-Amino-6-nitrotoluene, 3-Nitro-o-toluidine

  • CAS Number 603-83-8

  • Linear Formula CH3C6H3(NO2)NH2

  • Molecular Weight 152.15

  •  Beilstein Registry Number 388393

  •  EC Number 210-059-6

  •  MDL number MFCD00007731

  •  PubChem Substance ID 24846242

Purchase

Properties

InChI Key   HFCFJYRLBAANKN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
bp   305 °C(lit.)
mp   88-90 °C(lit.)
  89-92 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2660 6.1 / PGIII
WGK Germany 
3
RTECS 
XU8200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?