• USA Home
  • 09331 - 2-Methoxy-4-nitroaniline

EMAIL THIS PAGE TO A FRIEND
09331 Sigma-Aldrich

2-Methoxy-4-nitroaniline

purum, ≥99.0% (HPLC)

Synonym: 4-Nitro-o-anisidine

  • CAS Number 97-52-9

  • Linear Formula CH3OC6H3(NO2)NH2

  • Molecular Weight 168.15

  •  Beilstein Registry Number 879619

  •  EC Number 202-588-6

  •  MDL number MFCD00007363

  •  PubChem Substance ID 24846314

Purchase

Properties

InChI Key   GVBHRNIWBGTNQA-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
mp   139-143 °C
  140-142 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
BZ7170000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?