• USA Home
  • 09706 - Ammonium diethyldithiocarbamate

EMAIL THIS PAGE TO A FRIEND
09706 Sigma-Aldrich

Ammonium diethyldithiocarbamate

p.a.

Synonym: Diethyldithiocarbamic acid ammonium salt

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Building Blocks, Chemical Synthesis,
grade   p.a.
mp   80 °C (dec.) (lit.)
storage temp.   2-8°C
SMILES string   N.CCN(CC)C(S)=S
InChI   1S/C5H11NS2.H3N/c1-3-6(4-2)5(7)8;/h3-4H2,1-2H3,(H,7,8);1H3
InChI key   JSYCNFYVINPILE-UHFFFAOYSA-N

Description

Application

Reactant involved in:
• Reactions with copper acetylacetonato complex
• Triazole compounds
• Causing accelerated degradation of methyl iodide

Other Notes

Reagent used for the extraction of trace heavy metals, e.g. for AAS

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

359548 Ammonium diethyldithiocarbamate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?