EMAIL THIS PAGE TO A FRIEND
10440 Sigma-Aldrich

p-Anisaldehyde

purum, ≥98.0% (GC)

Synonym: 4-Methoxybenzaldehyde, Aubépine

  • CAS Number 123-11-5

  • Linear Formula CH3OC6H4CHO

  • Molecular Weight 136.15

  •  Beilstein Registry Number 471382

  •  EC Number 204-602-6

  •  MDL number MFCD00003385

Purchase

Properties

vapor density   4.7 (vs air)
InChI Key   ZRSNZINYAWTAHE-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.573(lit.)
  n20/D 1.573
bp   248 °C(lit.)
mp   −1 °C (lit.)
density   1.119 g/mL at 25 °C(lit.)

Description

Caution

may discolor to yellow on storage

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
BZ2625000
Flash Point(F) 
240.8 °F
Flash Point(C) 
116 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A88107 p-Anisaldehyde, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?