EMAIL THIS PAGE TO A FRIEND
10960 Sigma-Aldrich

Arbutin

≥96.0% (HPLC)

Synonym: 4-Hydroxyphenyl-β-D-glucopyranoside, p-Arbutin, Hydroquinone β-D-glucopyranoside

  • CAS Number 497-76-7

  • Empirical Formula (Hill Notation) C12H16O7

  • Molecular Weight 272.25

  •  Beilstein Registry Number 89673

  •  EC Number 207-850-3

  •  MDL number MFCD00016915

Purchase

Properties

InChI Key   BJRNKVDFDLYUGJ-RMPHRYRLSA-N
assay   ≥96.0% (HPLC)
optical activity   [α]20/D −65±2°, c = 2% in H2O
mp   195-198 °C
solubility   H2O: 50 mg/mL, clear (hot)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A4256 Arbutin, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?