• USA Home
  • 11390 - 5-Aza-2′-deoxycytidine

EMAIL THIS PAGE TO A FRIEND
11390 Sigma-Aldrich

5-Aza-2′-deoxycytidine

≥97.0% (HPLC)

Synonym: 2′-Deoxy-5-azacytidine, 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, Decitabine

  • CAS Number 2353-33-5

  • Empirical Formula (Hill Notation) C8H12N4O4

  • Molecular Weight 228.21

  •  Beilstein Registry Number 617982

  •  EC Number 219-089-4

  •  MDL number MFCD00043011

Purchase

Properties

InChI Key   XAUDJQYHKZQPEU-KVQBGUIXSA-N
assay   ≥97.0% (HPLC)
mp   ~200 °C (dec.)
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

A very potent cytotoxic agent to tumour cells in vitro at concentrations that do not inhibit macromolecular synthesis

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
XZ3012000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A3656 5-Aza-2′-deoxycytidine, ≥97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?