EMAIL THIS PAGE TO A FRIEND
11650 Supelco

Azulene

standard for GC, ≥99.0% (GC)

Synonym: Bicyclo[5.3.0]decapentaene

  • CAS Number 275-51-4

  • Empirical Formula (Hill Notation) C10H8

  • Molecular Weight 128.17

  •  Beilstein/REAXYS Number 969517

  •  EC Number 205-993-6

  •  MDL number MFCD00003810

  •  PubChem Substance ID 329749524

Purchase

Properties

grade   standard for GC
assay   ≥99.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   242 °C (lit.)
mp   98-100 °C (lit.)
Featured Industry   Environmental
format   neat
SMILES string   c1ccc2cccc2cc1
InChI   1S/C10H8/c1-2-5-9-7-4-8-10(9)6-3-1/h1-8H
InChI key   CUFNKYGDVFVPHO-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Quantity

λmax. 270 nm, log ε 4.72 in heptane

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
CO4570000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

37879 Azulene, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?