• USA Home
  • 13830 - 1-Benzyl-4-piperidone

EMAIL THIS PAGE TO A FRIEND
13830 Sigma-Aldrich

1-Benzyl-4-piperidone

technical, ≥98.0% (GC)

Synonym: 1-Benzyl-4-oxopiperidine

  • CAS Number 3612-20-2

  • Empirical Formula (Hill Notation) C12H15NO

  • Molecular Weight 189.25

  •  Beilstein/REAXYS Number 128556

  •  EC Number 222-782-4

  •  MDL number MFCD00006192

  •  PubChem Substance ID 329750358

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Piperidones
grade   technical
assay   ≥98.0% (GC)
refractive index   n20/D 1.541 (lit.)
  n20/D 1.541
bp   134 °C/7 mmHg (lit.)
density   1.021 g/mL at 25 °C (lit.)
SMILES string   O=C1CCN(CC1)Cc2ccccc2
InChI   1S/C12H15NO/c14-12-6-8-13(9-7-12)10-11-4-2-1-3-5-11/h1-5H,6-10H2
InChI key   SJZKULRDWHPHGG-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
159.8 °F
Flash Point(C) 
71 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B29806 1-Benzyl-4-piperidone, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?