EMAIL THIS PAGE TO A FRIEND
14390 Sigma-Aldrich

6-Biopterin

≥98.0% (sum of enantiomers, HPLC)

Synonym: (1’R,2’S)-Biopterin, 2-Amino-4-hydroxy-6-(1,2-dihydroxypropyl)pteridine, L-erythro-6-(1,2-Dihydroxypropyl)pterin

  • CAS Number 22150-76-1

  • Empirical Formula (Hill Notation) C9H11N5O3

  • Molecular Weight 237.22

  •  Beilstein Registry Number 87860

  •  EC Number 244-807-8

  •  MDL number MFCD00036787

Purchase

Properties

InChI Key   LHQIJBMDNUYRAM-DZSWIPIPSA-N
assay   ≥98.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −65±8°, c = 0.2% in 0.1 M HCl
storage temp.   2-8°C

Description

Other Notes

Review: Chemical mechanisms in the action of pteridine coenzymes

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
UO3506000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?