EMAIL THIS PAGE TO A FRIEND
15519 Sigma-Aldrich

1-Boc-4-piperidone

puriss., ≥99.0% (HPLC)

Synonym: N-Boc-4-piperidone, tert-Butyl 4-oxo-1-piperidinecarboxylate

  • CAS Number 79099-07-3

  • Empirical Formula (Hill Notation) C10H17NO3

  • Molecular Weight 199.25

  •  Beilstein/REAXYS Number 3650236

  •  MDL number MFCD00151800

  •  PubChem Substance ID 329750922

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Piperidones
InChI Key   ROUYFJUVMYHXFJ-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (HPLC)
mp   73-77 °C (lit.)
  73-77 °C

Description

Application

1-Boc-4-piperidone was used in the preparation of anti-aldol product with fair diastereo- and enantioselectivity.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?