EMAIL THIS PAGE TO A FRIEND
16250 Sigma-Aldrich

2-Bromoanisole

purum, ≥95.0% (GC)

Synonym: 1-Bromo-2-methoxybenzene

  • CAS Number 578-57-4

  • Linear Formula BrC6H4OCH3

  • Molecular Weight 187.03

  •  Beilstein Registry Number 1859996

  •  EC Number 209-425-8

  •  MDL number MFCD00000064

  •  PubChem Substance ID 24849985

Purchase

Properties

InChI Key   HTDQSWDEWGSAMN-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
contains   silver wool as stabilizer
refractive index   n20/D 1.573(lit.)
  n20/D 1.573
bp   223 °C(lit.)
mp   2 °C (lit.)
density   1.502 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
BZ8500000
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

159239 2-Bromoanisole, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?