EMAIL THIS PAGE TO A FRIEND
16255 Sigma-Aldrich

3-Bromoanisole

purum, ≥98.0% (GC)

Synonym: 1-Bromo-3-methoxybenzene

Purchase

Properties

InChI Key   PLDWAJLZAAHOGG-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
impurities   ≤0.5% 4-bromoanisole
refractive index   n20/D 1.563(lit.)
  n20/D 1.563
bp   210-211 °C(lit.)
density   1.490 g/mL at 20 °C
  1.477 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B56498 3-Bromoanisole, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?