EMAIL THIS PAGE TO A FRIEND
16280 Sigma-Aldrich

2-Bromobenzaldehyde

purum, ≥97.0% (GC)

Purchase

Properties

InChI Key   NDOPHXWIAZIXPR-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.595(lit.)
  n20/D 1.595
bp   230 °C(lit.)
mp   16-19 °C (lit.)
density   1.585 g/mL at 25 °C(lit.)

Description

Application

2-Bromobenzaldehyde was used in the synthesis of pentacyclic melodinus alkaloid (±)-meloscine and organomercurial compounds. It was also used as starting reagent during the asymmetric synthesis of a wide-spectrum agricultural fungicide, (S)-MA20565.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B57001 2-Bromobenzaldehyde, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?