EMAIL THIS PAGE TO A FRIEND
16340 Sigma-Aldrich

4-Bromobenzoic acid

puriss., ≥99.0% (HPLC)

  • CAS Number 586-76-5

  • Linear Formula BrC6H4CO2H

  • Molecular Weight 201.02

  •  Beilstein Registry Number 1906923

  •  EC Number 209-581-7

  •  MDL number MFCD00002529

  •  PubChem Substance ID 24849747

Purchase

Properties

InChI Key   TUXYZHVUPGXXQG-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (HPLC)
mp   252-254 °C(lit.)
  254-256 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
DG4448050

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?