EMAIL THIS PAGE TO A FRIEND
17320 Sigma-Aldrich

1-Bromohexane

purum, ≥98.0% (GC)

Synonym: Hexyl bromide

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
vapor density   5.7 (vs air)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.448 (lit.)
  n20/D 1.448
bp   154-158 °C (lit.)
mp   −85 °C (lit.)
density   1.176 g/mL at 25 °C (lit.)
SMILES string   CCCCCCBr
InChI   1S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3
InChI key   MNDIARAMWBIKFW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
RTECS 
MO0925000
Flash Point(F) 
134.6 °F
Flash Point(C) 
57 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?