EMAIL THIS PAGE TO A FRIEND
17366 Sigma-Aldrich

Neon

puriss., ≥99.99%

Purchase

Properties

vapor density   0.7 (21 °C, vs air)
grade   puriss.
assay   ≥99.99%
resistivity   6.97 μΩ-cm, 20°C
bp   −246 °C (lit.)
mp   -248.67 °C (lit.)
SMILES string   [Ne]
InChI   1S/Ne
InChI key   GKAOGPIIYCISHV-UHFFFAOYSA-N

Description

Packaging

pressure tin

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

Symbol 
GHS04  GHS04
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN1065 - class 2 - Neon, compressed
WGK Germany 
3
RTECS 
QP4450000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

control valve

Product #

Description

Add to Cart

97490 Flow controlling valve for 600 mL and 1000 mL pressure tins, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?