• USA Home
  • 17720 - 1-Bromo-4-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
17720 Sigma-Aldrich

1-Bromo-4-nitrobenzene

purum, ≥98.0% (HPLC)

  • CAS Number 586-78-7

  • Linear Formula BrC6H4NO2

  • Molecular Weight 202.01

  •  Beilstein Registry Number 636964

  •  EC Number 209-583-8

  •  MDL number MFCD00007280

  •  PubChem Substance ID 24850622

Purchase

Properties

InChI Key   ZDFBKZUDCQQKAC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
bp   255-256 °C(lit.)
mp   124-126 °C (lit.)
  124-126 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3459 6.1 / PGIII
WGK Germany 
3
RTECS 
CY9040550
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?