EMAIL THIS PAGE TO A FRIEND
17990 Sigma-Aldrich

2-Bromophenol

purum, ≥99.0% (HPLC)

  • CAS Number 95-56-7

  • Linear Formula BrC6H4OH

  • Molecular Weight 173.01

  •  Beilstein Registry Number 1905115

  •  EC Number 202-432-7

  •  MDL number MFCD00002146

  •  PubChem Substance ID 24850832

Purchase

Properties

InChI Key   VADKRMSMGWJZCF-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
refractive index   n20/D 1.589(lit.)
  n20/D 1.589
bp   195 °C(lit.)
mp   3-7 °C (lit.)
density   1.492 g/mL at 25 °C(lit.)
Gene Information   human ... ALOX12(239), ALOX15(246)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 3/PG 3
WGK Germany 
3
RTECS 
SJ7875000
Flash Point(F) 
107.6 °F
Flash Point(C) 
42 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?