EMAIL THIS PAGE TO A FRIEND
18010 Sigma-Aldrich

4-Bromophenol

purum, ≥98.0% (HPLC)

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   GZFGOTFRPZRKDS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
bp   235-236 °C(lit.)
mp   61-64 °C (lit.)
  61-65 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SJ7960000

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B75808 4-Bromophenol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?