• USA Home
  • 18039 - Holmium Standard for AAS

EMAIL THIS PAGE TO A FRIEND
18039 Supelco

Holmium Standard for AAS

TraceCERT®, 1000 mg/L Ho in nitric acid

Purchase

Properties

Related Categories AAS, AAS CRMs, Additional Standards, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
  TraceCERT®
shelf life   limited shelf life, expiry date on the label
concentration   1000 mg/L Ho in nitric acid
application(s)   AAS: suitable
format   single component solution

Description

Analysis Note

1000 mg/L Ho in 2% nitric acid, prepared with high purity Ho2O3, HNO3 and water

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?