• USA Home
  • 18087 - (±)-3,4-Methylenedioxymethamphetamine hydrochloride solution

EMAIL THIS PAGE TO A FRIEND
18087 Supelco

(±)-3,4-Methylenedioxymethamphetamine hydrochloride solution

~1 mg/mL in methanol, analytical standard, for drug analysis

Synonym: DL-3,4-Methylenedioxymethamphetamine hydrochloride solution, DL-MDMA hydrochloride solution, XTC hydrochloride solution

  • CAS Number 64057-70-1

  • Empirical Formula (Hill Notation) C11H15NO2 · HCl

  • Molecular Weight 229.70

  •  MDL number MFCD00079444

  •  PubChem Substance ID 329751619

Purchase

Properties

Related Categories Additional Standards, Amphetamine, Amphetamines / Stimulants Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard, for drug analysis
InChI Key   LUWHVONVCYWRMZ-UHFFFAOYSA-N
assay   ≥98.5% (HPLC)
shelf life   limited shelf life, expiry date on the label
drug control   USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)
concentration   ~1 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   corresponds for H-NMR
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   2-8°C

Description

General description

3,4-Methylenedioxymethamphetamine is considered as an illicit drug. It stimulates neurons to release serotonin and dopamine.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

ampoule contains 1.2mL solution

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Heroin, Cocaine, Amphetamine and MDMA on Ascentis® Express HILIC
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?