EMAIL THIS PAGE TO A FRIEND
22580 Sigma-Aldrich

D-(−)-Quinic acid

purum, ≥98.0% (T)

  • CAS Number 77-95-2

  • Empirical Formula (Hill Notation) C7H12O6

  • Molecular Weight 192.17

  •  Beilstein Registry Number 2212412

  •  EC Number 201-072-8

  •  MDL number MFCD00003864

  •  PubChem Substance ID 329752326

Purchase

Properties

Related Categories Allium cepa (Onion), Asymmetric Synthesis, Cell Biology, Chemical Synthesis, Chiral Resolution Reagents,
InChI Key   AAWZDTNXLSGCEK-WYWMIBKRSA-N
grade   purum
assay   ≥98.0% (T)
form   powder
optical activity   [α]20/D −44±1°, c = 20% in H2O
ign. residue   ≤0.1%
mp   165-170 °C

Description

Other Notes

Use as chiral building block; and references cited therein.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GU8650000

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?