EMAIL THIS PAGE TO A FRIEND
23360 Sigma-Aldrich

Chloranilic acid

puriss. p.a., for the determination of Ca, Sr, Zr, ≥99.0% (T)

Synonym: 2,5-Dichloro-3,6-dihydroxy-p-benzoquinone, 2,5-Dichloro-3,6-dihydroxy-2,5-cyclohexadiene-1,4-dione, NSC 6108, NSC 97383

  • CAS Number 87-88-7

  • Empirical Formula (Hill Notation) C6H2Cl2O4

  • Molecular Weight 208.98

  •  Beilstein Registry Number 1875040

  •  EC Number 201-780-7

  •  MDL number MFCD00001596

  •  PubChem Substance ID 329752414

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Building Blocks, C-D, Puriss p.a.,
InChI Key   IPPWILKGXFOXHO-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.0% (T)
quality   for the determination of Ca, Sr, Zr
mp   ≥300 °C(lit.)

Description

Other Notes

Reagent for the paperchromatographic detection of metals

Application

Reactant involved in:
• Acting as a proton donor in reactions studying dimensionality control
• Synthesis of dimethylbipyridyl complexes
• Synthesis of (nonylbenzimidazolylmethyl)benzene for preparation of neutral altitudinal rotor-shaped dirhenium metallacycles
• Charge-transfer reactions with metoprolol tartrate
• Salt formation with organic bases
• Synthesis of osmium metalla-rectangles with anticancer activity

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
DK4005000

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?