• USA Home
  • 23720 - 4-Chlorobenzotrifluoride

EMAIL THIS PAGE TO A FRIEND
23720 Sigma-Aldrich

4-Chlorobenzotrifluoride

purum, ≥98.0% (GC)

Synonym: 1-Chloro-4-(trifluoromethyl)benzene, 4-Chloro-α,α,α-trifluorotoluene

  • CAS Number 98-56-6

  • Linear Formula ClC6H4CF3

  • Molecular Weight 180.55

  •  Beilstein Registry Number 510203

  •  EC Number 202-681-1

  •  MDL number MFCD00000627

  •  PubChem Substance ID 24854225

Purchase

Properties

InChI Key   QULYNCCPRWKEMF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.446(lit.)
  n20/D 1.447
bp   136-138 °C(lit.)
mp   −36 °C (lit.)
density   1.353 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2234 3 / PGIII
WGK Germany 
2
RTECS 
XS9145000
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?