• USA Home
  • 27640 - O-Phosphorylethanolamine

EMAIL THIS PAGE TO A FRIEND
27640 Sigma-Aldrich

O-Phosphorylethanolamine

≥99.0% (T)

Synonym: 2-Aminoethyl dihydrogen phosphate, O-Phosphocolamine, O-Phosphoethanolamine, Colaminephosphoric acid

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   SUHOOTKUPISOBE-UHFFFAOYSA-N
assay   ≥99.0% (T)
mp   235-240 °C
storage temp.   −20°C

Description

Other Notes

Potent inhibitor of ornithine decarboxylase

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
KJ5830000

Documents

Certificate of Analysis


Expore Enzymes, Proteins & Cofactors

Biomedical Applications
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P0503 O-Phosphorylethanolamine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?