• USA Home
  • 28570 - 2-Pyridinecarbonitrile

EMAIL THIS PAGE TO A FRIEND
28570 Sigma-Aldrich

2-Pyridinecarbonitrile

purum, ≥98.0% (GC)

Synonym: 2-Cyanopyridine, Picolinonitrile

  • CAS Number 100-70-9

  • Empirical Formula (Hill Notation) C6H4N2

  • Molecular Weight 104.11

  •  Beilstein/REAXYS Number 107710

  •  EC Number 202-880-3

  •  MDL number MFCD00006218

  •  PubChem Substance ID 329753240

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.529 (lit.)
bp   212-215 °C (lit.)
mp   24-27 °C (lit.)
  24-27 °C
density   1.081 g/mL at 25 °C (lit.)
storage temp.   2-8°C
SMILES string   N#Cc1ccccn1
InChI   1S/C6H4N2/c7-5-6-3-1-2-4-8-6/h1-4H
InChI key   FFNVQNRYTPFDDP-UHFFFAOYSA-N

Description

Caution

may discolor to light-brown on storage

Other Notes

Precursor of the appropriate amidate for the modification of proteins via amidation

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
192.2 °F
Flash Point(C) 
89 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?