• USA Home
  • 28590 - 4-Pyridinecarbonitrile

EMAIL THIS PAGE TO A FRIEND
28590 Sigma-Aldrich

4-Pyridinecarbonitrile

purum, ≥96.0% (GC)

Synonym: 4-Cyanopyridine, Isonicotinic acid nitrile, Isonicotinonitrile

  • CAS Number 100-48-1

  • Empirical Formula (Hill Notation) C6H4N2

  • Molecular Weight 104.11

  •  Beilstein/REAXYS Number 107712

  •  EC Number 202-856-2

  •  MDL number MFCD00006417

  •  PubChem Substance ID 329753253

Purchase

Properties

Related Categories Building Blocks, C6, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
grade   purum
assay   ≥96.0% (GC)
mp   76-79 °C (lit.)
  77-81 °C
SMILES string   N#Cc1ccncc1
InChI   1S/C6H4N2/c7-5-6-1-3-8-4-2-6/h1-4H
InChI key   GPHQHTOMRSGBNZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C95005 4-Pyridinecarbonitrile, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?