• USA Home
  • 29890 - Cyclopropanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
29890 Sigma-Aldrich

Cyclopropanecarboxylic acid

purum, ≥98.0% (T)

Synonym: CPC-acid

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
grade   purum
assay   ≥98.0% (T)
refractive index   n20/D 1.438 (lit.)
  n20/D 1.438
bp   182-184 °C (lit.)
mp   14-17 °C (lit.)
density   1.081 g/mL at 25 °C (lit.)
SMILES string   OC(=O)C1CC1
InChI   1S/C4H6O2/c5-4(6)3-1-2-3/h3H,1-2H2,(H,5,6)
InChI key   YMGUBTXCNDTFJI-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
RTECS 
GZ1100000
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C116602 Cyclopropanecarboxylic acid, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?