• USA Home
  • 30372 - 6-FAM-phosphoramidite

EMAIL THIS PAGE TO A FRIEND
30372 Sigma-Aldrich

6-FAM-phosphoramidite

≥95.0% (HPLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Probes, Labels, Particles and Stains, Other Fluorescent Conjugates
InChI Key   MGPYJVWEJNTXLC-UHFFFAOYSA-N
assay   ≥95.0% (HPLC)
suitability   corresponds for fluorescence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?