• USA Home
  • 31070 - 2′-Deoxyguanosine monohydrate

EMAIL THIS PAGE TO A FRIEND
31070 Sigma-Aldrich

2′-Deoxyguanosine monohydrate

≥98.0% (HPLC)

Synonym: 9-(2-Deoxy-β-D-ribofuranosyl)guanine, Guanine-2′-deoxyriboside

  • CAS Number 312693-72-4

  • Empirical Formula (Hill Notation) C10H13N5O4 · H2O

  • Molecular Weight 285.26

  •  Beilstein/REAXYS Number 39814

  •  EC Number 213-505-8

  •  MDL number MFCD00150760

  •  PubChem Substance ID 329753638

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleosides, Nucleotides, Oligonucleotides More...
assay   ≥98.0% (HPLC)
solubility   1 M NH4OH: 50 mg/mL, clear to very faintly turbid, yellow to brown
SMILES string   O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1
InChI   1S/C10H13N5O4.H2O/c11-10-13-8-7(9(18)14-10)12-3-15(8)6-1-4(17)5(2-16)19-6;/h3-6,16-17H,1-2H2,(H3,11,13,14,18);1H2/t4-,5+,6+;/m0./s1
InChI key   LZSCQUCOIRGCEJ-FPKZOZHISA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Deoxyribonucleosides on Ascentis® Si

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Deoxyribonucleosides on Ascentis® Si
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Nucleosides on Ascentis® Si (Silica)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Nucleosides on Ascentis® Si (Silica)
Keywords: Chromatography, High performance liquid chromatography

UHPLC/MS Analysis of 8-Hydroxy-2′-Deoxyguanosine and Related Compounds on Titan™ Silica

From our library of Articles, Sigma-Aldrich presents UHPLC/MS Analysis of 8-Hydroxy-2′-Deoxyguanosine and Related Compounds on Titan™ Silica
Keywords: Chromatography, Mass spectrometry, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?