EMAIL THIS PAGE TO A FRIEND
31574 Sigma-Aldrich

Silafluofen

PESTANAL®, analytical standard

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Insecticides, Other Insecticides,
grade   analytical standard
InChI Key   HPYNBECUCCGGPA-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?